Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triethyl 2-fluoro-2-phosphonoacetate, 96%

Catalog No. AAL1791303
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1791303 1 g
AAL1791306 5 g
AAL1791314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL1791303 Supplier Thermo Scientific Chemicals Supplier No. L1791303
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 2356-16-3 | C8H16FO5P | 242.183 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2356-16-3
Molecular Formula C8H16FO5P
Molecular Weight (g/mol) 242.183
MDL Number MFCD00134400
InChI Key FVPISMANESAJQZ-UHFFFAOYSA-N
Synonym triethyl 2-fluoro-2-phosphonoacetate, 2-fluoro-2-phosphonoacetic acid triethyl ester, ethyl 2-diethoxyphosphoryl-2-fluoroacetate, triethyl phosphonofluoroacetate, ethyl diethoxyphosphoryl fluoro acetate, triethyl-2-fluoro-2-phosphonoacetate, acetic acid, diethoxyphosphinyl fluoro-, ethyl ester, 2-fluoro-2-phosphonoacetic acidtriethylester, diethyl ethoxycarbonyl fluoromethyl phosphonate, diethoxyphosphinyl fluoroacetic acid ethyl ester
PubChem CID 2734865
IUPAC Name ethyl 2-diethoxyphosphoryl-2-fluoroacetate
SMILES CCOC(=O)C(F)P(=O)(OCC)OCC

Specifications

Density 1.194
Boiling Point 75°C (0.01 mmHg)
Flash Point 73°C (163°F)
Refractive Index 1.425
Quantity 1 g
Beilstein 1912161
Formula Weight 242.19
Percent Purity 96%
Chemical Name or Material Triethyl 2-fluoro-2-phosphonoacetate
Hazard Category H227-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.