Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Triethyl 3-Methyl-4-Phosphono-2-Butenoate, Mixture Of Cis And Trans

Catalog No. 3389585G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
33-895-85G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 33-895-85G Supplier Sigma Aldrich Supplier No. 3389585G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Reactant for preparation of:Bicyclodecenes and oxabicyclodecenes via intramolecular [4 + 3]-cycloaddition, using alkylation, olefination and couplingMethyloctatrienoic acid retinoid X receptor modulators from insulin sensitizers for the treatment of type II diabetes

Specifications

CAS 6033-23-4
Density 0.815 g/mL at 25°C
Boiling Point 149°C to 150°C
Molecular Formula C7H16O
Linear Formula CH3(CH2)4CH(OH)CH3
Refractive Index n20/D 1.46
Quantity 5 g
MDL Number MFCD00065003
Synonym Diethyl 3-ethoxycarbonyl-2-methyl-2-propenyl phosphonate; Triethyl 3-methyl-4-phosphonocrotonate; Triethyl 4-phosphonosenecioate, mixture of cis and trans
Molecular Weight (g/mol) 116.2 g/mol
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.