Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Triethyl 4-Phosphonocrotonate, Mixture Of Isomers

Catalog No. 11609210G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-609-210G 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Warning
Catalog No. 11-609-210G Supplier Sigma Aldrich Supplier No. 11609210G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Reactant for: IminodIpr∗

Specifications

CAS 10034-76-1
Molecular Formula CaO4S -+ 0.5H2O
Refractive Index n20/D 1.455
Linear Formula CaSO4 -+ 0.5H2O
MDL Number MFCD00799067
Quantity 1 kg
Synonym 4-(Diethoxyphosphinyl)-2-butenoic acid ethyl ester; 4-Phosphonocrotonic acid triethyl ester
Molecular Weight (g/mol) 145.15 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.