Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Triethyl Borate

Catalog No. T59307100ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T59307100ML 100 mL
T59307500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. T59307100ML Supplier Sigma Aldrich Supplier No. T59307100ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Triethyl borate may be used as boron source to synthesize lithium borophosphates, alumina borosilicate ceramic nanofibres and boron-doped single wall carbon nanotubes.

Specifications

CAS 112-27-6
Melting Point -7°C
Density 1.124 g/mL at 20°C
Boiling Point 125°C to 127°C (0.1 mmHg, lit.)
Molecular Formula C6H14O4
Refractive Index n20/D 1.374
Linear Formula HO(CH2CH2O)2CH2CH2OH
MDL Number MFCD00002880
Quantity 100 mL
Synonym Boric acid triethyl ester; Boron ethoxide; Triethoxyborane
Molecular Weight (g/mol) 150.17 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.