Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triethyl borate, 97%, AcroSeal™, Thermo Scientific Chemicals

Catalog No. AC428658000
Change view
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC428658000 800 mL AcroSeal™ Glass Bottle
AC428651000 100 mL AcroSeal™ Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC428658000 Supplier Thermo Scientific Chemicals Supplier No. 428658000
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 150-46-9 | Molecular Formula: C6H15BO3 | Molecular Weight: 145.99 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 150-46-9
Molecular Formula C6H15BO3
Molecular Weight (g/mol) 145.99
MDL Number MFCD00009073
InChI Key AJSTXXYNEIHPMD-UHFFFAOYSA-N
Synonym triethoxyborane, boron ethoxide, boron triethoxide, triethoxyboron, boric acid, triethyl ester, boric acid triethyl ester, borane, triethoxy, triethylborate, ethyl borate eto 3b, boric acid h3bo3 , triethyl ester
PubChem CID 9009
ChEBI CHEBI:38916
IUPAC Name triethyl borate
SMILES B(OCC)(OCC)OCC

Specifications

Melting Point -84.5°C
Density 0.8580g/mL
Boiling Point 117.0°C to 118.0°C
Flash Point 11°C
Assay Percent Range 96% min. (GC)
Packaging AcroSeal™ Glass Bottle
Refractive Index 1.3720 to 1.3740
Linear Formula (C2H5O)3B
Quantity 800 mL
Specific Gravity 0.858
Solubility Information Solubility in water: decomposes
Formula Weight 145.99
Percent Purity 97%
Chemical Name or Material Triethyl borate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 205-760-9

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.