Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Triethyl citrate

Catalog No. 275001LF Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
27-500-1LF 1 L
27-500-250MLF 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 27-500-1LF Supplier Sigma Aldrich Supplier No. 275001LF
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Triethyl citrate is a safe direct food additive.Triethyl citrate was used as a plasticizer in the study of the feasibility of near-infrared (NIR) spectroscopy in determining the influence of plasticizer levels and sintering temperature on the breaking force of extended-release matrix tablets prepared by roller-compaction.

Specifications

CAS 77-93-0
Density 1.14 g/mL (at 25°C (literature))
Boiling Point 235°C (150 mmHg)
Molecular Formula C12H20O7
Linear Formula HOC(COOC2H5)(CH2COOC2H5)2
Refractive Index n20/D 1.442 (literature)
MDL Number MFCD00009201
Quantity 1 L
Synonym Ethyl citrate
Molecular Weight (g/mol) 276.28
Percent Purity ≥98.0% (GC)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.