Learn More
Medchemexpress LLC Triethyl citrate | 77-93-0 | 122584 | 98.9% | 276.29 | 5 G
SDP
Catalog No. 5000453053
Encompass_Preferred
Triethyl citrate is an orally active citrate compound that can be used in research on plasticizers for food packaging materials. It induces acute toxic effects such as weakness, depression, ataxia, hyperexcitability, restlessness, dripping urine, and irregular, labored breathing. This product is for research use only and is not sold to patients.
  • Orally active citrate compound.
  • Can be utilized in research for plasticizers for food packaging materials.
  • Available in liquid form.
Catalog No. 50-004-53053 Supplier Medchemexpress LLC Supplier No. HYW0116025G
Triethyl citrate is an orally active citrate compound that can be used in research on plasticizers for food packaging materials. It induces acute toxic effects such as weakness, depression, ataxia, hyperexcitability, restlessness, dripping urine, and irregular, labored breathing. This product is for research use only and is not sold to patients.
  • Orally active citrate compound.
  • Can be utilized in research for plasticizers for food packaging materials.
  • Available in liquid form.