Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triethyl Citrate, FCC, 99-100.5%, Spectrum™ Chemical
SDP

Catalog No. 18607567 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
500 mL
4 L
5 gal.
200 L
Packaging:
Amber Glass Bottle
Drum
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-607-567 500 mL Amber Glass Bottle
18-607-568 4 L Amber Glass Bottle
18-607-569 200 L Drum
18-607-570 5 gal. Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-607-567 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. T1171500MLGL
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

T1171, 77-93-0, C12H20O7

Spectrum™ Chemical Triethyl Citrate, FCC is a colorless, odorless liquid used as a food additive (E number E1505) to stabilize foams, especially as whipping aid for egg whites. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage, nutritional supplements and personal care applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 77-93-0
Molecular Formula C12H20O7
Molecular Weight (g/mol) 276.29
InChI Key DOOTYTYQINUNNV-UHFFFAOYSA-N
IUPAC Name 1,2,3-triethyl 2-hydroxypropane-1,2,3-tricarboxylate
SMILES CCOC(=O)CC(O)(CC(=O)OCC)C(=O)OCC

Specifications

CAS Max % 1
Packaging Amber Glass Bottle
Refractive Index 1.439-1.443
Quantity 500 mL
Percent Purity 99.0 to 100.5%
Grade FCC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.