Learn More
U.S. Pharmacopeia Triethyl Citrate | 77-93-0 | MFCD00009201 | 100 mg
Catalog No. NC9408961
Encompass
Molecular formula C12-H20-O7 | Molecular weight 276.29 | Melting Point -67 °F (-55 °C) | Boiling Point 561.2 °F (294 °C) | Synonyms: Ethyl citrate
Catalog No. NC9408961 Supplier U.S. Pharmacopeia Supplier No. 1683606
Molecular formula C12-H20-O7 | Molecular weight 276.29 | Melting Point -67 °F (-55 °C) | Boiling Point 561.2 °F (294 °C) | Synonyms: Ethyl citrate