Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triethyl O-Acetylcitrate 97.0+%, TCI America™
SDP

Catalog No. A0086500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A0086500G 500 g
A008625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A0086500G Supplier TCI America Supplier No. A0086500G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 77-89-4 | C14H22O8 | 318.322 g/mol

Chemical Identifiers

CAS 77-89-4
Molecular Formula C14H22O8
Molecular Weight (g/mol) 318.322
MDL Number MFCD00049378
InChI Key WEAPVABOECTMGR-UHFFFAOYSA-N
Synonym acetyl triethyl citrate, triethyl o-acetylcitrate, citroflex a 2, acetyltriethyl citrate, atec, triethyl acetylcitrate, triethyl 2-acetoxypropane-1,2,3-tricarboxylate, o-acetylcitric acid triethyl ester, citric acid, acetyl triethyl ester, 1,2,3-propanetricarboxylic acid, 2-acetyloxy-, triethyl ester
PubChem CID 6504
IUPAC Name triethyl 2-acetyloxypropane-1,2,3-tricarboxylate
SMILES CCOC(=O)CC(CC(=O)OCC)(C(=O)OCC)OC(=O)C

Specifications

Color Yellow
Boiling Point 229°C
Quantity 500 g
Formula Weight 318.32
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Triethyl O-Acetylcitrate
EINECSNumber (2)-1327
RTECSNumber GE8225000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.