Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Triethyl orthopropionate

Catalog No. T60607100ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T60607100ML 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T60607100ML Supplier Sigma Aldrich Supplier No. T60607100ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Triethyl orthopropionate

Specifications

CAS 115-80-0
Density 0.876 g/mL (at 25°C (literature))
Boiling Point 155°C to 160°C (lit.)
Molecular Formula C9H20O3
Linear Formula CH3CH2C(OC2H5)3
Refractive Index n20/D 1.402 (literature)
MDL Number MFCD00009226
Quantity 100 g
Synonym 1,1,1-Triethoxypropane
Molecular Weight (g/mol) 176.25
Percent Purity 97%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.