Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Triethyl phosphate

Catalog No. 5387284L Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 mL
1 L
4 L
18 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
53-872-84L 4 L
53-872-8100ML 100 mL
53-872-818L 18 L
53-872-81L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 53-872-84L Supplier Sigma Aldrich Supplier No. 5387284L
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Triethyl phosphate is an alkyl phosphate
  • It participates in the thermal transformation of sodium acetate-2-c14 to n-butyl acetate-2-c14.tandem mass spectrometry data independently generated by scripps center for metabolomics is available to view or download in pdf
  • 538728.pdf tested metabolites are featured on scripps center for metabolomics metlin metabolite database
  • To learn more, visit sigma.com/metlin.reagentplus is a registered trademark of sigma-aldrich co
  • Llc

Specifications

CAS 78-40-0
Density 1.072 g/mL (at 25°C (literature))
Boiling Point 215°C (lit.)
Molecular Formula C6H15O4P
Refractive Index n20/D 1.403 (literature)
Linear Formula (C2H5O)3 PO
Quantity 4 L
MDL Number MFCD00009077
Synonym TEP; Ethyl phosphate
Molecular Weight (g/mol) 182.15
Percent Purity ≥99.8%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.