Learn More
Medchemexpress LLC Triethyl phosphate | 78-40-0 | MFCD00009077 | 100.0% | 182.15 g/mol | C6H15O4P | 500 G
SDP
Catalog No. 5000302329
Encompass_Preferred
Triethyl phosphate is an organophosphorus solvent and plasticizer used in chemical synthesis and materials applications. It is supplied as a high-purity reagent suitable for use as a reaction medium, solvent, or additive in polymer formulations for laboratory and research purposes.
  • High purity reagent suitable for analytical and synthetic use.
  • Acts as an effective solvent for a variety of organic reactions.
  • Functions as a plasticizer to improve polymer flexibility and durability.
  • Low volatility and thermal stability for elevated-temperature processes.
  • Useful as an analytical marker in environmental studies.
Catalog No. 50-003-02329 Supplier Medchemexpress LLC Supplier No. HYW016414500G
Triethyl phosphate is an organophosphorus solvent and plasticizer used in chemical synthesis and materials applications. It is supplied as a high-purity reagent suitable for use as a reaction medium, solvent, or additive in polymer formulations for laboratory and research purposes.
  • High purity reagent suitable for analytical and synthetic use.
  • Acts as an effective solvent for a variety of organic reactions.
  • Functions as a plasticizer to improve polymer flexibility and durability.
  • Low volatility and thermal stability for elevated-temperature processes.
  • Useful as an analytical marker in environmental studies.