Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triethyl phosphate, 98+%

Catalog No. aaa1793836
Change view
Click to view available options
Quantity:
500 g
2500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1793836 500 g
AAA179380E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1793836 Supplier Thermo Scientific Chemicals Supplier No. A1793836
Only null left
Add to Cart

CAS: 78-40-0 | C6H15O4P | 182.16 g/mol

Triethyl phosphate finds it major applications in plastics industry as a flame retardant, plasticizer and carrier, where it is available in the matrix. A further 10 to 20 % are used in other industrial branches as a solvent, plasticizer, flame retardant or intermediate for the production of pharmaceuticals, and lacquers. Triethyl phosphate is a useful synthetic intermediate used in the synthesis of mesoporous spheres of metal oxides and phosphates. It is also used as an industrial catalyst employed in ketene synthesis where the compound is hydrolyzed, as a polymer resin modifier, as a solvent, a car paint repairing product and as flame retarder

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Triethyl phosphate finds it major applications in plastics industry as a flame retardant, plasticizer and carrier, where it is available in the matrix. A further 10 to 20 % are used in other industrial branches as a solvent, plasticizer, flame retardant or intermediate for the production of pharmaceuticals, and lacquers. Triethyl phosphate is a useful synthetic intermediate used in the synthesis of mesoporous spheres of metal oxides and phosphates. It is also used as an industrial catalyst employed in ketene synthesis where the compound is hydrolyzed, as a polymer resin modifier, as a solvent, a car paint repairing product and as flame retarder

Solubility
Fully miscible in water at 20°C.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78-40-0
Molecular Formula C6H15O4P
Molecular Weight (g/mol) 182.16
MDL Number MFCD00009077
InChI Key DQWPFSLDHJDLRL-UHFFFAOYSA-N
Synonym triethylphosphate, phosphoric acid, triethyl ester, tris ethyl phosphate, triethoxyphosphine oxide, triethylfosfat, ethyl phosphate eto 3po, triethylfosfat czech, ethyl phosphate van, unii-qih4k96k7j, phosphoric acid triethyl ester
PubChem CID 6535
ChEBI CHEBI:45927
IUPAC Name triethyl phosphate
SMILES CCOP(=O)(OCC)OCC

Specifications

Density 1.072
Boiling Point 215°C
Flash Point 115°C (240°F)
Odor Mild
Refractive Index 1.405
Quantity 500 g
Merck Index 14,9674
Solubility Information Fully miscible in water at 20°C.
Formula Weight 182.16
Percent Purity ≥98%
Chemical Name or Material Triethyl phosphate
Show More Show Less
Hazard Category H302
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 201-114-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.