Alfa Aesar™ Triethyl phosphate, 99+%

Manufacturer: Alfa Aesar™ 4000114

Quantity 25g

 View more versions of this product

Catalog No. AA4000114

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 78-40-0
    Formula Weight 182.16
    MDL Number MFCD00009077
    Merck Index 14,9674
    Chemical Name or Material Triethyl phosphate

    Specifications

    Formula Weight 182.16
    Health Hazard 3 GHS P Statement
    P264-P270-P301+P312-P330-P501a
    Wash thoroughly after handling.
    Do not eat, drink or smoke when using this product.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Rinse mouth.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    MDL Number MFCD00009077
    Health Hazard 2 GHS H Statement
    H302
    Harmful if swallowed.
    Density 1.072g/mL
    Boiling Point 215°C
    Refractive Index 1.4050
    Physical Form Liquid
    Merck Index 14,9674
    Flash Point 115° (240°F)
    TSCA TSCA
    Odor Mild
    CAS 78-40-0
    Health Hazard 1 Warning
    Quantity 25g
    Chemical Name or Material Triethyl phosphate
    Assay ≥99%

    GHS H Statement
    H302
    Harmful if swallowed.

    GHS P Statement
    P264-P270-P301+P312-P330-P501a
    Wash thoroughly after handling.
    Do not eat, drink or smoke when using this product.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Rinse mouth.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit