Learn More
Medchemexpress LLC Triethyl phosphate-d15 | 135942-11-9 | MFCD08705044 | 99.6% | 197.25 g/mol | C6D15O4P | 100 MG
SDP
Catalog No. 5000302078
Encompass_Preferred
Triethyl phosphate-d15 is the deuterium-labeled analogue of triethyl phosphate, provided as a stable isotope tracer and internal standard for analytical and research applications. It is intended for use in mass spectrometry and metabolic studies to improve quantitation, identification, and method validation where isotopically labeled references are required.
  • Deuterium-labeled internal standard for accurate mass spectrometry quantitation.
  • High isotopic and chemical purity for reliable analytical performance.
  • Suitable for use as a tracer in metabolic and pharmacokinetic studies.
  • Compatible with GC-MS and LC-MS workflows.
  • Stable when stored under recommended conditions (protect from light; refrigerated or frozen in solvent).
  • Available in small milligram quantities for research and method development.
Catalog No. 50-003-02078 Supplier Medchemexpress LLC Supplier No. HYW016414S100MG
Triethyl phosphate-d15 is the deuterium-labeled analogue of triethyl phosphate, provided as a stable isotope tracer and internal standard for analytical and research applications. It is intended for use in mass spectrometry and metabolic studies to improve quantitation, identification, and method validation where isotopically labeled references are required.
  • Deuterium-labeled internal standard for accurate mass spectrometry quantitation.
  • High isotopic and chemical purity for reliable analytical performance.
  • Suitable for use as a tracer in metabolic and pharmacokinetic studies.
  • Compatible with GC-MS and LC-MS workflows.
  • Stable when stored under recommended conditions (protect from light; refrigerated or frozen in solvent).
  • Available in small milligram quantities for research and method development.