Learn More
Medchemexpress LLC Triethyl phosphate-d15 | 135942-11-9 | MFCD08705044 | 99.6% | 197.25 g/mol | C6D15O4P | 25 MG
SDP
Catalog No. 5000303718
Encompass_Preferred
Triethyl phosphate-d15 is the deuterium-labeled isotopologue of triethyl phosphate used as an analytical standard and tracer for LC-MS and other mass spectrometry applications. Supplied as a high-purity reference material for quantitative analysis and method development (research use only).
  • Deuterium-labeled internal standard for LC-MS quantification.
  • High isotopic enrichment, reducing interference in mass spectra.
  • High chemical purity suitable for analytical standards.
  • Available in small milligram pack sizes for trace-level work.
  • Stable when stored under recommended conditions, preserving integrity.
Catalog No. 50-003-03718 Supplier Medchemexpress LLC Supplier No. 25MGHYW016414S
Triethyl phosphate-d15 is the deuterium-labeled isotopologue of triethyl phosphate used as an analytical standard and tracer for LC-MS and other mass spectrometry applications. Supplied as a high-purity reference material for quantitative analysis and method development (research use only).
  • Deuterium-labeled internal standard for LC-MS quantification.
  • High isotopic enrichment, reducing interference in mass spectra.
  • High chemical purity suitable for analytical standards.
  • Available in small milligram pack sizes for trace-level work.
  • Stable when stored under recommended conditions, preserving integrity.