Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Triethyl Phosphite

Catalog No. T612045ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 mL
100 mL
500 mL
4 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T612045ML 5 mL
T61204500ML 500 mL
T612044L 4 L
T61204100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Warning
Catalog No. T612045ML Supplier Sigma Aldrich Supplier No. T612045ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Used as a reducing agent; can react with electrophiles to form phosphonates or phosphates; forms a stable complex with copper(I) iodide.

Specifications

CAS 122-52-1
Density 0.969 g/mL at 25°C
Boiling Point 156°C
Molecular Formula C6H15O3P
Refractive Index n20/D 1.413
Linear Formula (C2H5O)3P
MDL Number MFCD00009084
Quantity 5 mL
Synonym P(EtO)3; P(OEt)3; P(EtO)3; P(OEt)3
Molecular Weight (g/mol) 166.16 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.