Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Triethylaluminum Solution

Catalog No. 252662800ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
800 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
25-266-2800ML 800 mL
25-266-2100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 25-266-2800ML Supplier Sigma Aldrich Supplier No. 252662800ML
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Triethylaluminum solution can be used: As a reagent in Alumino-Mannich reaction, where it reacts with iminium ion substrates for which the corresponding Petasis borono-Mannich reaction is unsuccessful.In the synthesis of macrocyclic binuclear aluminum complexes with efficient activities toward ring-opening polymerization of +a-caprolactone.To promote Claisen rearrangement of allyl vinyl ether derivatives along with the transfer of ethyl group to aldehydic carbon.As a reagent for copper-catalyzed asymmetric conjugate addition of alkenyl- and alkylalanes to +a,+a-unsaturated lactams.As a precursor to synthesize diethylaluminium cyanide, which is used as hydrocyanating reagent.

Specifications

CAS 97-93-8
Density 0.692 g/mL at 25°C
Molecular Formula C6H15Al
Linear Formula Al(C2H5)3
MDL Number MFCD00009015
Quantity 800 mL
Synonym Aluminumtriethanide
Molecular Weight (g/mol) 114.16 g/mol
Biological Activity Central Nervous System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.