Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triethylamine Borane 90.0+%, TCI America™
SDP

Catalog No. T118025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T118025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T118025G Supplier TCI America Supplier No. T118025G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1722-26-5
Molecular Formula C6H18BN
MDL Number MFCD00012423
Synonym Borane - Triethylamine Complex, TEAB

Specifications

Color Yellow
Boiling Point 100°C
Quantity 25 g
UN Number 2924
Formula Weight 115.03
Percent Purity ≥90.0% (T)
Physical Form Liquid
Chemical Name or Material Triethylamine Borane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.