Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triethylamine hydrochloride, 98%

Catalog No. AAA173180B
Change view
Click to view available options
Quantity:
250 g
1000 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA173180B 1000 g
AAA1731830 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA173180B Supplier Thermo Scientific Chemicals Supplier No. A173180B
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 554-68-7 | C6H16ClN | 137.65 g/mol

It finds its application as a protein purification reagent. Triethylamine is used as a competing base for the separation of acidic basic and neutral drugs by reverse-phased high-performance liquid chromatography (HPLC). Triethylamine induces visual disturbances (such as foggy vision) in humans, and is also used in industry as a quenching agent in the ozonolysis of alkenes (e.g. (E)-2-Pentene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It finds its application as a protein purification reagent. Triethylamine is used as a competing base for the separation of acidic basic and neutral drugs by reverse-phased high-performance liquid chromatography (HPLC). Triethylamine induces visual disturbances (such as foggy vision) in humans, and is also used in industry as a quenching agent in the ozonolysis of alkenes (e.g. (E)-2-Pentene.

Solubility
Soluble in water at 20°C 1440 g/L. Soluble in ethanol, chloroform, and alcohol. Insoluble in ether.

Notes
Hygroscopic. Store away from water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas. Protect from humidity.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 554-68-7
Molecular Formula C6H16ClN
Molecular Weight (g/mol) 137.65
MDL Number MFCD00012500
InChI Key ILWRPSCZWQJDMK-UHFFFAOYSA-N
Synonym triethylamine hydrochloride, triethylammonium chloride, triethylamine hcl, ethanamine, n,n-diethyl-, hydrochloride, triethylamine monohydrochloride, triethylamine, hydrochloride, unii-0nx3818gcw, amine, triethyl, hydrochloride, n,n-diethylethanamine hydrochloride, triethylaminehydrochloride
PubChem CID 11130
IUPAC Name N,N-diethylethanamine;hydrochloride
SMILES [H+].[Cl-].CCN(CC)CC

Specifications

Melting Point ∼260°C (decomposition)
Density 1.07
pH 5
Odor Amine-like
Linear Formula (CH3CH2)3N·HCl
Quantity 1000 g
Beilstein 3906409
Sensitivity Hygroscopic
Solubility Information Soluble in water at 20°C 1440g/L. Soluble in ethanol,chloroform,and alcohol. Insoluble in ether.
Formula Weight 137.65
Percent Purity 98%
Chemical Name or Material Triethylamine hydrochloride
Show More Show Less
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 209-067-2
RTECSNumber YE2111500
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.