Learn More
Boston Bioproducts Inc Triethylammonium Acetate (1 M, pH 7.0) - 500ml
Catalog No. NC2402406
Encompass
The Triethylammonium Acetate (TEAA) buffer is a versatile buffer used in biological applications. With a pH range between 6.9-7.1, it serves as an ion-pairing reagent for chromatographic separations, especially in oligonucleotide research. TEAA also has applications in proteomics studies, glycopeptide separation, and liquid chromatography-mass spectrometry (LC-MS) analyses of peptides and proteins. It has also been used for the purification of chemically synthesized nucleotides and DNA.
Catalog No. NC2402406 Supplier Boston Bioproducts Inc Supplier No. BB2820500ML
The Triethylammonium Acetate (TEAA) buffer is a versatile buffer used in biological applications. With a pH range between 6.9-7.1, it serves as an ion-pairing reagent for chromatographic separations, especially in oligonucleotide research. TEAA also has applications in proteomics studies, glycopeptide separation, and liquid chromatography-mass spectrometry (LC-MS) analyses of peptides and proteins. It has also been used for the purification of chemically synthesized nucleotides and DNA.