Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triethylborane (ca. 11% in Tetrahydrofuran, ca. 1mol/L), TCI America™
SDP

Catalog No. T1984100ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T1984100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T1984100ML Supplier TCI America Supplier No. T1984100ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 97-94-9
Molecular Formula C6H15B
Molecular Weight (g/mol) 98.00
MDL Number MFCD00009022
InChI Key LALRXNPLTWZJIJ-UHFFFAOYSA-N
Synonym triethylboron, borane, triethyl, triethylborine, boron triethyl, boron ethyl, unii-z3s980z4p3, hsdb 897, triethylboran, borethyl, triethyl borane
PubChem CID 7357
IUPAC Name triethylborane
SMILES CCB(CC)CC

Specifications

Color Colorless
Quantity 100 mL
UN Number 2924
Formula Weight 98.00
Physical Form Liquid
Chemical Name or Material Triethylborane (ca. 11% in Tetrahydrofuran, ca. 1mol/L)
EINECSNumber (2)-2978
RTECSNumber ED2100000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.