Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Triethylborane Solution

Catalog No. 179701800ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
800 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
17-970-1800ML 800 mL
17-970-1100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 17-970-1800ML Supplier Sigma Aldrich Supplier No. 179701800ML
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Triethylborane is used as a catalyst for:Allylation of aldehydesDecarboxylative C-C bond cleavage reactionsRhenium hydride / boron Lewis acid cocatalysis of alkene hydrogenationsRegioselective hydroxyalkylation of unsaturated oxime ethersIt can be employed as a reactant for radical reductions of alkyl bromides with N-heterocyclic carbene boranes and for the synthesis of tetramethylammonium trialkylphenylborate salts.

Specifications

CAS 97-94-9
Density 0.865 g/mL at 25°C
Molecular Formula C6H15B
Linear Formula (C2H5)3B
MDL Number MFCD00009022
Quantity 800 mL
Molecular Weight (g/mol) 97.99 g/mol
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.