Learn More
Strem, An Ascensus Company CAS# 97-94-9. 100g. Triethylborane, 98%. MFCD00009022
SDP
Catalog No. 5090113741
Encompass
CAS# 97-94-9. 100g. Triethylborane, 98%. MFCD00009022. Molecular Weight: 98.00. Molecular Formula: B(C2H5)3. Color/form: colorless liq. Strem# 93-0540. http://www.strem.com/catalog/v/93-0540/
Catalog No. 50-901-13741 Supplier Strem, An Ascensus Company Supplier No. 930540100G
CAS# 97-94-9. 100g. Triethylborane, 98%. MFCD00009022. Molecular Weight: 98.00. Molecular Formula: B(C2H5)3. Color/form: colorless liq. Strem# 93-0540. http://www.strem.com/catalog/v/93-0540/