Learn More
Medchemexpress LLC Triethylene glycol | 112-27-6 | MFCD00002880 | 150.17 g·mol⁻1 | C6H14O4 | 100 MG
SDP
Catalog No. 5000300357
Encompass_Preferred
Triethylene glycol (CAS 112-27-6) is an analytical standard used as a reference material for research and analytical method development. It is a colorless to light-yellow liquid with formula C6H14O4 and molecular weight 150.17 g·mol⁻1, supplied in measured small-quantity packs for calibration and validation in analytical workflows.
  • Analytical standard for calibration and method validation.
  • Suitable for chromatography and mass spectrometry applications.
  • High purity with documentation; COA and SDS available.
  • Supplied in precise small quantities for accurate standard preparation.
  • Colorless to light-yellow liquid; stable when stored per instructions.
Catalog No. 50-003-00357 Supplier Medchemexpress LLC Supplier No. HYW017440R100MG
Triethylene glycol (CAS 112-27-6) is an analytical standard used as a reference material for research and analytical method development. It is a colorless to light-yellow liquid with formula C6H14O4 and molecular weight 150.17 g·mol⁻1, supplied in measured small-quantity packs for calibration and validation in analytical workflows.
  • Analytical standard for calibration and method validation.
  • Suitable for chromatography and mass spectrometry applications.
  • High purity with documentation; COA and SDS available.
  • Supplied in precise small quantities for accurate standard preparation.
  • Colorless to light-yellow liquid; stable when stored per instructions.