Learn More
Medchemexpress LLC Triethylene glycol | 112-27-6 | MFCD00002880 | >=99.0% | 150.17 g/mol | C6H14O4 | 500 MG
SDP
Catalog No. 5000301779
Encompass_Preferred
Triethylene glycol (CAS 112-27-6) is a low-molecular-weight polyethylene glycol used as an analytical standard and a PEG-based linker for research and synthesis. It has molecular formula C6H14O4 and molecular weight 150.17 g/mol, and is supplied at high purity for analytical and synthetic workflows.
  • High purity suitable for analytical methods (≥99.0% by GC).
  • Suitable as a PEG-based linker in small-molecule synthesis.
  • Useful for LC, GC, and MS method development and calibration.
  • Low molecular weight with polar solvent properties for formulation and synthesis.
  • Supplied with datasheet information for safe handling and storage.
Catalog No. 50-003-01779 Supplier Medchemexpress LLC Supplier No. HYW017440R500MG
Triethylene glycol (CAS 112-27-6) is a low-molecular-weight polyethylene glycol used as an analytical standard and a PEG-based linker for research and synthesis. It has molecular formula C6H14O4 and molecular weight 150.17 g/mol, and is supplied at high purity for analytical and synthetic workflows.
  • High purity suitable for analytical methods (≥99.0% by GC).
  • Suitable as a PEG-based linker in small-molecule synthesis.
  • Useful for LC, GC, and MS method development and calibration.
  • Low molecular weight with polar solvent properties for formulation and synthesis.
  • Supplied with datasheet information for safe handling and storage.