Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Triethylene glycol | 112-27-6 | MFCD00002880 | 99.9% | 150.17 g/mol | C6H14O4 | 1 KG
SDP

Catalog No. 5000301754
Encompass_Preferred

Triethylene glycol (CAS 112-27-6) is a short polyethylene glycol (PEG) diol used as a flexible spacer and linker in small-molecule conjugation and PROTAC synthesis. Supplied as a high-purity liquid, it is suitable for synthesis workflows and formulation.

  • High purity suitable for research synthesis.
  • Liquid form with density 1.124 g/cm3 for easy handling.
  • Soluble in water and PBS at 100 mg/mL; ultrasonic assistance recommended.
  • Molecular weight 150.17 g/mol and formula C6H14O4.
  • Stable under recommended storage: pure form -20°C (3 years) and 4°C (2 years).

Catalog No. 50-003-01754 Supplier Medchemexpress LLC Supplier No. HYW0174401KG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Triethylene glycol (CAS 112-27-6) is a short polyethylene glycol (PEG) diol used as a flexible spacer and linker in small-molecule conjugation and PROTAC synthesis. Supplied as a high-purity liquid, it is suitable for synthesis workflows and formulation.

  • High purity suitable for research synthesis.
  • Liquid form with density 1.124 g/cm3 for easy handling.
  • Soluble in water and PBS at 100 mg/mL; ultrasonic assistance recommended.
  • Molecular weight 150.17 g/mol and formula C6H14O4.
  • Stable under recommended storage: pure form -20°C (3 years) and 4°C (2 years).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.