Learn More
Medchemexpress LLC Triethylene glycol | 112-27-6 | MFCD00002880 | 99.9% | 150.17 g/mol | C6H14O4 | 500 G
SDP
Catalog No. 5000300478
Encompass_Preferred
Triethylene glycol (TEG) is a PEG-based linker reagent used in chemical synthesis, including PROTAC assembly and other linker chemistries. Supplied as a high-purity liquid, it is intended for research use and for incorporation into synthetic workflows requiring a short polyethylene glycol spacer.
  • High purity (99.88%) suitable for synthesis applications.
  • Useful as a PEG-based linker for PROTAC and other conjugation chemistry.
  • Liquid, colorless to light yellow, easy to measure and dispense.
  • Molecular weight 150.17 g/mol for accurate formulation and stoichiometry.
  • Available in larger pack sizes to support scale-up.
  • Stable when stored under recommended conditions for extended shelf life.
Catalog No. 50-003-00478 Supplier Medchemexpress LLC Supplier No. HYW017440500G
Triethylene glycol (TEG) is a PEG-based linker reagent used in chemical synthesis, including PROTAC assembly and other linker chemistries. Supplied as a high-purity liquid, it is intended for research use and for incorporation into synthetic workflows requiring a short polyethylene glycol spacer.
  • High purity (99.88%) suitable for synthesis applications.
  • Useful as a PEG-based linker for PROTAC and other conjugation chemistry.
  • Liquid, colorless to light yellow, easy to measure and dispense.
  • Molecular weight 150.17 g/mol for accurate formulation and stoichiometry.
  • Available in larger pack sizes to support scale-up.
  • Stable when stored under recommended conditions for extended shelf life.