Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Triethylene Glycol

Catalog No. T5945520KG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 g
1 kg
20 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T5945520KG 20 kg
T594551KG 1 kg
T5945525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T5945520KG Supplier Sigma Aldrich Supplier No. T5945520KG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Triethylene Glycol

Specifications

CAS 112-27-6
Melting Point -7°C
Density 1.124 g/mL at 20°C
Boiling Point 125°C to 127°C (0.1 mmHg, lit.)
Molecular Formula C6H14O4
Refractive Index n20/D 1.455
Linear Formula HO(CH2CH2O)2CH2CH2OH
MDL Number MFCD00002880
Quantity 20 kg
Synonym Triglycol
Molecular Weight (g/mol) 150.17 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.