Learn More
Medchemexpress LLC Triethylene glycol bis(p-toluenesulfonate) | 19249-03-7 | 99.9% | 458.54 | 100 G
SDP
Catalog No. 5000454704
Encompass_Preferred
Triethylene glycol bis(p-toluenesulfonate) is a PEG-based PROTAC linker. It can be utilized in the synthesis of PROTACs, which are molecules containing two different ligands connected by a linker. One ligand is for an E3 ubiquitin ligase and the other is for the target protein, allowing PROTACs to exploit the intracellular ubiquitin-proteasome system for selective degradation of target proteins.
  • Can be used in the synthesis of PROTACs.
  • PROTACs utilize the intracellular ubiquitin-proteasome system.
  • Enables selective degradation of target proteins.
Catalog No. 50-004-54704 Supplier Medchemexpress LLC Supplier No. HYW013731100G
Triethylene glycol bis(p-toluenesulfonate) is a PEG-based PROTAC linker. It can be utilized in the synthesis of PROTACs, which are molecules containing two different ligands connected by a linker. One ligand is for an E3 ubiquitin ligase and the other is for the target protein, allowing PROTACs to exploit the intracellular ubiquitin-proteasome system for selective degradation of target proteins.
  • Can be used in the synthesis of PROTACs.
  • PROTACs utilize the intracellular ubiquitin-proteasome system.
  • Enables selective degradation of target proteins.