Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Triethylene glycol dimethacrylate

Catalog No. 2615481L Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
26-154-81L 1 L
26-154-8250ML 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 26-154-81L Supplier Sigma Aldrich Supplier No. 2615481L
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Triethylene glycol dimethacrylate (TEGDMA) is a hydrophilic, low viscosity, difunctional methacrylic monomer employed as a crosslinking agent.TEGDMA is a monomer typically used in dental resin materials that can cause specific stress responses in eukaryotic cells.

Specifications

CAS 109-16-0
Density 1.092 g/mL (at 25°C)
Boiling Point 170°C to 172°C (5 mmHg)
Molecular Formula C14H22O6
Refractive Index n20/D 1.461 (literature)
Linear Formula CH2=C(CH3)COO(CH2CH2O)3 COC(CH3)=CH2
MDL Number MFCD00008591
Quantity 1 L
Molecular Weight (g/mol) 286.32
Percent Purity 95%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.