Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Triethylene glycol mono-11-mercaptoundecyl ether

Catalog No. 673110250MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
67-311-0250MG 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 67-311-0250MG Supplier Sigma Aldrich Supplier No. 673110250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Triethylene glycol mono-11-mercaptoundecyl ether is an alkanethiol that forms a self-assembled monolayer (SAM) on a variety of surfaces.Triethylene glycol mono-11-mercaptoundecyl ether can be used in the formation of low cost micro-patterns for biomedical and microelectronics based applications. It can also be used as a SAM for surface immobilization of various molecular species on gold electrodes for biological applications.

Specifications

CAS 130727-41-2
Density 0.995 g/mL (at 25°C)
Molecular Formula C17H36O4S
Refractive Index n20/D 1.476
Linear Formula HS(CH2)11(OCH2CH2)3 OH
MDL Number MFCD08457662
Quantity 250 mg
Synonym (11-Mercaptoundecyl)tri(ethylene glycol)
Molecular Weight (g/mol) 336.53
Percent Purity 95%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.