Learn More
Medchemexpress LLC Triethylene glycol monobenzyl ether | 55489-58-2 | MFCD02093556 | 99.8% | 240.30 g/mol | C13H20O4 | 10 G
SDP
Catalog No. 5000210203
Encompass_Preferred
Triethylene glycol monobenzyl ether is a polyethylene glycol (PEG)-based PROTAC linker used in the synthesis of proteolysis-targeting chimeras (PROTACs). It is a colorless to light yellow liquid with a molecular weight of 240.30 g/mol and CAS number 55489-58-2.
  • PEG-based linker suitable for PROTAC synthesis
  • High purity suitable for research applications
  • Liquid form enables facile coupling and handling
  • Commonly supplied in small laboratory package sizes for synthetic use
Catalog No. 50-002-10203 Supplier Medchemexpress LLC Supplier No. HYW04445910G
Triethylene glycol monobenzyl ether is a polyethylene glycol (PEG)-based PROTAC linker used in the synthesis of proteolysis-targeting chimeras (PROTACs). It is a colorless to light yellow liquid with a molecular weight of 240.30 g/mol and CAS number 55489-58-2.
  • PEG-based linker suitable for PROTAC synthesis
  • High purity suitable for research applications
  • Liquid form enables facile coupling and handling
  • Commonly supplied in small laboratory package sizes for synthetic use