Learn More
Medchemexpress LLC Triethylene glycol monobenzyl ether | 55489-58-2 | MFCD02093556 | 99.8% | 240.30 g/mol | C13H20O4 | 25 G
SDP
Catalog No. 5000209324
Encompass_Preferred
Triethylene glycol monobenzyl ether is a PEG-based PROTAC linker used as a flexible spacer to connect ligands in proteolysis-targeting chimera (PROTAC) synthesis. It is supplied for research use with analytical documentation available.
  • High purity: 99.8% (HPLC).
  • PEG-based linker suitable for PROTAC synthesis.
  • Molecular formula: C13H20O4; molecular weight: 240.30 g/mol.
  • Available in laboratory pack sizes, including 25 g.
  • Provided with datasheet, COA, and SDS for research use.
Catalog No. 50-002-09324 Supplier Medchemexpress LLC Supplier No. 25GHYW044459
Triethylene glycol monobenzyl ether is a PEG-based PROTAC linker used as a flexible spacer to connect ligands in proteolysis-targeting chimera (PROTAC) synthesis. It is supplied for research use with analytical documentation available.
  • High purity: 99.8% (HPLC).
  • PEG-based linker suitable for PROTAC synthesis.
  • Molecular formula: C13H20O4; molecular weight: 240.30 g/mol.
  • Available in laboratory pack sizes, including 25 g.
  • Provided with datasheet, COA, and SDS for research use.