Learn More
Medchemexpress LLC Triethylene glycol monododecyl ether | 3055-94-5 | MFCD00042659 | 100.0% | 318.49 | C18H38O4 | 50 MG
SDP
Catalog No. 5000283043
Encompass_Preferred
Triethylene glycol monododecyl ether is a PEG-based PROTAC linker combining a triethylene glycol spacer with a dodecyl (C12) alkyl tail, used as a flexible spacer in the synthesis of PROTAC molecules. The product is supplied with manufacturer documentation including a datasheet, SDS, and COA, and is listed with high purity and defined molecular properties.
  • PEG-based triethylene glycol spacer for linker design.
  • Dodecyl alkyl tail for hydrophobic anchoring.
  • High listed purity (99.98%).
  • Molecular weight 318.49 and formula C18H38O4.
  • Documentation available: datasheet, SDS, and COA.
  • Suitable as a PROTAC linker in small-molecule synthesis.
Catalog No. 50-002-83043 Supplier Medchemexpress LLC Supplier No. HYW14250650MG
Triethylene glycol monododecyl ether is a PEG-based PROTAC linker combining a triethylene glycol spacer with a dodecyl (C12) alkyl tail, used as a flexible spacer in the synthesis of PROTAC molecules. The product is supplied with manufacturer documentation including a datasheet, SDS, and COA, and is listed with high purity and defined molecular properties.
  • PEG-based triethylene glycol spacer for linker design.
  • Dodecyl alkyl tail for hydrophobic anchoring.
  • High listed purity (99.98%).
  • Molecular weight 318.49 and formula C18H38O4.
  • Documentation available: datasheet, SDS, and COA.
  • Suitable as a PROTAC linker in small-molecule synthesis.