Learn More
Medchemexpress LLC Triethylene glycol monododecyl ether | 3055-94-5 | MFCD00042659 | 100.0% | 318.49 g/mol | C18H38O4 | 250 MG
SDP
Catalog No. 5000283543
Encompass_Preferred
Triethylene glycol monododecyl ether is a polyethylene glycol-based linker used in the synthesis of proteolysis targeting chimeras (PROTACs). It provides a flexible, hydrophilic spacer that can improve solubility and help control the distance between binding ligands, supporting linker optimization during medicinal chemistry and chemical biology research.
  • Provides a flexible, hydrophilic spacer for bifunctional molecules.
  • Improves aqueous solubility of conjugated ligands.
  • Enables modulation of inter-ligand distance in PROTAC design.
  • Compatible with common organic synthesis transformations.
  • Supplied in small-scale quantities suitable for research and development.
Catalog No. 50-002-83543 Supplier Medchemexpress LLC Supplier No. HYW142506250MG
Triethylene glycol monododecyl ether is a polyethylene glycol-based linker used in the synthesis of proteolysis targeting chimeras (PROTACs). It provides a flexible, hydrophilic spacer that can improve solubility and help control the distance between binding ligands, supporting linker optimization during medicinal chemistry and chemical biology research.
  • Provides a flexible, hydrophilic spacer for bifunctional molecules.
  • Improves aqueous solubility of conjugated ligands.
  • Enables modulation of inter-ligand distance in PROTAC design.
  • Compatible with common organic synthesis transformations.
  • Supplied in small-scale quantities suitable for research and development.