Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triethylene Glycol Monododecyl Ether 95.0+%, TCI America™
SDP

Catalog No. T07685ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T07685ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T07685ML Supplier TCI America Supplier No. T07685ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3055-94-5
Molecular Formula C18H38O4
Molecular Weight (g/mol) 318.50
MDL Number MFCD00042659
InChI Key FKMHSNTVILORFA-UHFFFAOYSA-N
PubChem CID 76458
IUPAC Name 2-{2-[2-(dodecyloxy)ethoxy]ethoxy}ethan-1-ol
SMILES CCCCCCCCCCCCOCCOCCOCCO

Specifications

Color Colorless
Quantity 5 mL
Formula Weight 318.50
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Triethylene Glycol Monododecyl Ether
EINECSNumber (7)-0097
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.