Learn More
Strem, An Ascensus Company CAS# 4730-54-5. 25MG. 1,4,7-Triazacyclononane, 97%. MFCD00012358
SDP
Supplier:  Strem, An Ascensus Company 07250025MG
Encompass
CAS# 4730-54-5. 25MG. 1,4,7-Triazacyclononane, 97%. MFCD00012358. Molecular Formula: C6H15N3. Molecular Weight: 129.20. Color/Form: white xtl. Strem# 07-2500. www.strem.com/catalog/v3/07-2500/
Catalog No. 50-196-3623