Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Triethylgermanium hydride

Catalog No. 4299611G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
42-996-11G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 42-996-11G Supplier Sigma Aldrich Supplier No. 4299611G
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Triethylgermanium hydride (Ge(C2H5)3H) may be prepared by a three step process:Ge(C2H5)3Br reacted with Na to yield Ge2(C2H5)6Ge2(C2H5)6 treated with Li to form Ge(C2H5)3Liammonolysis of Ge(C2H5)3Li to give the end product, Ge(C2H5)3H. 1Reagent undergoes hydrogermylation with alkyl propiolates to give triethylgermylacrylates.

Specifications

CAS 1188-14-3
Density 0.994 g/mL (at 25°C)
Boiling Point 121°C to 122°C (20 mmHg)
Molecular Formula C6H16Ge
Linear Formula (C2H5)3 GeH
Refractive Index n20/D 1.433 (literature)
Quantity 1 g
MDL Number MFCD00077989
Synonym Triethylgermane
Molecular Weight (g/mol) 160.83
Percent Purity 98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.