Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Triethyloxonium Tetrafluoroborate

Catalog No. 90520100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mg
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
90-520-100G 25 g
90-520-25G 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 90-520-100G Supplier Sigma Aldrich Supplier No. 90520100G
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Triethyloxonium tetrafluoroborate can be used: To prepare amino esters by reacting with lactams followed by hydrolysis.In the preparation of substituted imidazolines from aziridinesandnitriles via [3+2]-cycloaddition reaction.For the N-alkylation of a series of N-arylsulfonyl-+a-amino acid methyl esters having variable substituents at 4th position of the sulfonamide aromatic ring.Powerful ethylating agent; Esterification of acids; Modifies carboxyl residues in proteins

Specifications

CAS 1345974-62-0
Molecular Formula C6H15BF4O
Linear Formula (C2H5)3O(BF4)
MDL Number MFCD00044423
Quantity 25 g
Molecular Weight (g/mol) 189.99 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.