Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sigma Aldrich Triethyloxonium Tetrafluoroborate
Shop All MilliporeSigma Sigma Organic Chemistry Products
Description
Triethyloxonium tetrafluoroborate can be used: To prepare amino esters by reacting with lactams followed by hydrolysis.In the preparation of substituted imidazolines from aziridinesandnitriles via [3+2]-cycloaddition reaction.For the N-alkylation of a series of N-arylsulfonyl-+a-amino acid methyl esters having variable substituents at 4th position of the sulfonamide aromatic ring.Powerful ethylating agent; Esterification of acids; Modifies carboxyl residues in proteins
Specifications
Specifications
| CAS | 1345974-62-0 |
| Melting Point | >300°C |
| Molecular Formula | C88H64Br4O8Rh2 |
| Linear Formula | C88H64Br4O8Rh2 |
| Quantity | 250 mg |
| Synonym | Davies dirhodium catalyst; Dirhodium tetrakis((S,R)-1-(4-bromophenyl)-2,2-diphenylcyclopropanecarboxylate); Dirhodium tetrakis[(R)-1-(4-bromophenyl)-2,2- diphenylcyclopropane carboxylate]; Tetrakis[(R)-(-)-[(1R)-1-(4-bromophenyl)-2,2-diphenylcyclopropanecarboxylato]dirhodium(II); Tetrakis[(R)-1-(4-bromophenyl)-2,2- diphenylcyclopropane carboxylato]dirhodium(II) |
| Molecular Weight (g/mol) | 1774.87 g/mol |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?