Learn More
Strem, An Ascensus Company CAS# 78-40-0. 500g. Triethylphosphate, 99%. MFCD00009077
SDP
Catalog No. 5090114230
Encompass
CAS# 78-40-0. 500g. Triethylphosphate, 99%. MFCD00009077. Molecular Weight: 182.16. Molecular Formula: (C2H5O)3PO. Color/form: colorless liq. Strem# 93-1559. http://www.strem.com/catalog/v/93-1559/
Catalog No. 50-901-14230 Supplier Strem, An Ascensus Company Supplier No. 931559500G
CAS# 78-40-0. 500g. Triethylphosphate, 99%. MFCD00009077. Molecular Weight: 182.16. Molecular Formula: (C2H5O)3PO. Color/form: colorless liq. Strem# 93-1559. http://www.strem.com/catalog/v/93-1559/