Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Triethylphosphine

Catalog No. 2452755G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
24-527-55G 5 g
24-527-525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 24-527-55G Supplier Sigma Aldrich Supplier No. 2452755G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Triethylphosphine is generally used as a ligand in the organometallic chemistry. It can be used in:Synthesis of the tetrahedrally coordinated L3Fe-Nx complex with a terminal nitride group.Synthesis of (PEt3)2Ni(++2-C14H10), a source of the reactive Ni(PEt3)2 moiety.Synthesis of dinuclear rhodium complexes with triethyllphosphane bridges.This container should be opened only by a technically qualified person.

Specifications

CAS 554-70-1
Density 0.802 g/mL at 20°C
Boiling Point 127°C to 128°C
Molecular Formula C6H15P
Refractive Index n20/D 1.456
Linear Formula (C2H5)3P
MDL Number MFCD00009256
Quantity 5 g
Molecular Weight (g/mol) 118.16 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.