Learn More
Strem, An Ascensus Company CAS# 554-70-1. 25g. Triethylphosphine, 99%. MFCD00009256
SDP
Catalog No. 502045257
Encompass
CAS# 554-70-1. 25g. Triethylphosphine, 99%. MFCD00009256. Molecular Weight: 118.16. Molecular Formula: (C2H5)3P. Color/form: colorless liq. Strem# 15-6300. http://www.strem.com/catalog/v/15-6300/
Catalog No. 50-204-5257 Supplier Strem, An Ascensus Company Supplier No. 15630025G
CAS# 554-70-1. 25g. Triethylphosphine, 99%. MFCD00009256. Molecular Weight: 118.16. Molecular Formula: (C2H5)3P. Color/form: colorless liq. Strem# 15-6300. http://www.strem.com/catalog/v/15-6300/