Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Triethylphosphine Solution

Catalog No. 90144025ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
90-144-025ML 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 90-144-025ML Supplier Sigma Aldrich Supplier No. 90144025ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Offered as the 2-methyltetrahydrofuran solution for more convenient handling, triethylphosphine is a valuable ligand for:Regioselective formation of alcohols from hydrocarbonylation of alkenes.Bronsted acid-assisted synthesis of dihydropyrones from aromatic aldehydes and allenoates.Silaboration of enynes.Conversion of phenylacetic acids to phenylacetonitriles of benzonitriles.Thermal decomposition of platinum and palladium naphthalenediyl.Asymmetrical Morita-Baylis-Hillman reaction.

Specifications

CAS 1366421-67-1
Molecular Formula C36H55N3O -+ HCl
Linear Formula C36H55N3O -+ HCl
Quantity 1 g
Synonym PEt3; Triethylphosphorous solution
Molecular Weight (g/mol) 582.3 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.