Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Triethylphosphine Solution

Catalog No. 34617950ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
34-617-950ML 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 34-617-950ML Supplier Sigma Aldrich Supplier No. 34617950ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Catalyst for: Regioselective formation of alcohols from hydrocarbonylation of alkenesBronsted acid-assisted synthesis of dihydropyrones from aromatic aldehydes and allenoatesSilaboration of enynesConversion of phenylacetic acids to phenylacetonitriles of benzonitrilesThermal decomposition of platinum and palladium naphthalenediylAsymmetrical Morita-Baylis-Hillman reaction

Specifications

CAS 617-55-0
Density 1.223 g/mL at 25°C
Molecular Formula C6H10O5
Linear Formula CH3O2CCH2CH(OH)CO2CH3
MDL Number MFCD00066215
Quantity 5 g
Synonym PEt3; Triethylphosphorous solution
Molecular Weight (g/mol) 162.14 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.