Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Triethylsilane

Catalog No. 467448100ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
46-744-8100ML 100 mL
46-744-825ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 46-744-8100ML Supplier Sigma Aldrich Supplier No. 467448100ML
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Triethylsilane can be used as:A reducing agent in the regioselective reductive coupling of enones and allenes.A reagent in the redox initiated cationic polymerization.A reagent in catalytic transfer hydrogenation, reduction of alkyl halides and silylation of aromatic C-H bonds, etc.As a reagent for the generation of indium hydride (Cl2InH) applicable as a catalyst for the intramolecular cyclization of enynes.

Specifications

CAS 617-86-7
Density 0.728 g/mL at 25°C
Boiling Point 107°C to 108°C
Molecular Formula C6H16Si
Refractive Index n20/D 1.412
Linear Formula (C2H5)3SiH
MDL Number MFCD00009018
Quantity 100 mL
Synonym NSC 93579; Triethylhydrosilane; Triethylsilicon hydride
Molecular Weight (g/mol) 116.28 g/mol
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.