Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triethylsilane, 99%

Catalog No. p-4509581
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Glass bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC212920050 5 g Glass bottle
AC212920250 25 g Glass bottle
AC212921000 100 g Glass bottle
AC212925000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. AC212920050 Supplier Thermo Scientific Chemicals Supplier No. 212920050
Only null left
Add to Cart
This item is not returnable. View return policy

CAS: 617-86-7 | C6H16Si | 116.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 617-86-7
Molecular Formula C6H16Si
Molecular Weight (g/mol) 116.28
InChI Key QXTIBZLKQPJVII-UHFFFAOYSA-N
Synonym triethylsilane, hydride, silane, triethyl, triethylhydrosilane, triethylsilyl, silane e3h, c6h16si, triethylsilyl radical, silane, triethyl-,, triethylsilane tes
PubChem CID 6327258
IUPAC Name triethylsilicon
SMILES CC[Si](CC)CC

Specifications

Boiling Point 107.0°C to 108.0°C
Color Colorless
Physical Form Liquid
Quantity 5 g
Assay Percent Range 99%
Linear Formula (C2H5)3SiH
Beilstein 04,625
Flash Point −3°C
Fieser 01,1218; 02,433; 03,304; 04,530; 05,694; 12,482; 13,340; 14,322; 15,338; 16,356; 17,367
Merck Index 15,9841
Solubility Information Solubility in water: decomposes
Formula Weight 116.28
Percent Purity 99%
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.4100 to 1.4120
Specific Gravity 0.72
Density 0.7200g/mL
Chemical Name or Material Triethylsilane
Show More Show Less
Hazard Category GHS Signal Word: Danger
EINECSNumber 210-535-3

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.