Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triethylsilanol, 97%

Catalog No. p-7043966
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0373803 1 g
AAL0373806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0373803 Supplier Thermo Scientific Chemicals Supplier No. L0373803
Only null left
Add to Cart

CAS: 597-52-4 | C6H16OSi | 132.278 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 597-52-4
Molecular Formula C6H16OSi
Molecular Weight (g/mol) 132.278
MDL Number MFCD00042640
InChI Key WVMSIBFANXCZKT-UHFFFAOYSA-N
Synonym triethylsilanol, silanol, triethyl, hydroxytriethylsilane, silanol, 1,1,1-triethyl, triethyl hydroxy silane, triethyl hydroxy silicon, tesoh, c2h5 3sioh, acmc-1an47
PubChem CID 69005
IUPAC Name triethyl(hydroxy)silane
SMILES CC[Si](CC)(CC)O

Specifications

Boiling Point 154°C
Quantity 1 g
UN Number UN1993
Beilstein 1732848
Flash Point 57°C (135°F)
Formula Weight 132.28
Percent Purity 97%
Odor Mild
Refractive Index 1.433
Density 0.86
Chemical Name or Material Triethylsilanol
Show More Show Less
Hazard Category H226
Hazard Statement GHS H Statement
H226
Flammable liquid and vapor.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 209-903-6
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.